Gene Transcripts

Gene Symbol
Trim30b
Entrez Gene ID
244183
Gene Name
tripartite motif-containing 30B
Gene Aliases
A530023O14Rik, Trim30-1
Chromosome Location
Chr. 7: 104355398 - 104358646 on Build GRCm38
UniGene ID
Mm.347647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175648.2 NP_783579.1 2 698
GenBank mRNA AK041488.1 - 2 698
AK162946.1 - 3132

View Full Product Details