Gene Transcripts

Gene Symbol
Anapc5
Entrez Gene ID
59008
Gene Name
anaphase-promoting complex subunit 5
Gene Aliases
2510006G12Rik, AA408751, AA536819, AA986414, APC5, Anpc5
Chromosome Location
Chr. 5: 122787459 - 122821342 on Build GRCm38
UniGene ID
Mm.332667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042491.2 NP_001035956.1 15 1960
NM_001289517.1 NP_001276446.1 15 1960
NM_001289518.1 NP_001276447.1 15 1999
NM_001289519.1 NP_001276448.1 15 1979
NM_001289520.1 NP_001276449.1 15 1975
NM_021505.3 NP_067480.2 15 1999
GenBank mRNA AB041593.1 - 15 1943
AK012579.1 - 8 899
AK031432.1 - 15 1924
AK049719.1 - 15 1953
AK075969.1 - 15 1916
AK088327.1 - 15 1962
AK148679.1 - 15 1915
AK151314.1 - 15 1906
AK151905.1 - 15 1945
AK152528.1 - 15 1875
AK152981.1 - 15 1907
AK153373.1 - 15 1906
AK153422.1 - 15 1906
AK159204.1 - 15 1906
AK159713.1 - 15 1914
AK159916.1 - 15 1938
AK167561.1 - 14 2875
AK168135.1 - 15 1923
AK168737.1 - 15 1900
AK168814.1 - 15 1938
AK169196.1 - 15 1962
AK201518.1 - 2 147
AK212170.1 - 2 147
BC010339.1 - 15 1915 2648173
BC011386.1 - 2 132
BC046566.1 - 15 1939 2650790
BC046804.1 - 15 1897 6404923

View Full Product Details