Gene Transcripts
- Gene Symbol
- Btnl6
- Entrez Gene ID
- 624681
- Gene Name
- butyrophilin-like 6
- Gene Aliases
- EG624681, Gm6519, NG13
- Chromosome Location
- Chr. 17: 34507935 - 34517352 on Build GRCm38
- UniGene ID
- Mm.471592
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030747.1 |
NP_109672.1
|
4 |
866 |
|
View Full Product Details