Gene Transcripts

Gene Symbol
Tmub1
Entrez Gene ID
64295
Gene Name
transmembrane and ubiquitin-like domain containing 1
Gene Aliases
2010004O20Rik, AB030183, Hops
Chromosome Location
Chr. 5: 24445463 - 24448331 on Build GRCm38
UniGene ID
Mm.150098
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022418.3 NP_071863.1 2 435
GenBank mRNA AB030183.1 - 2 397
AK008109.1 - 2 369
AY603379.1 - 2 366
BC019547.1 - 2 391 4164765

View Full Product Details