Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026549.3 | NP_080825.1 | 3 | 307 | |
GenBank mRNA | AK003339.1 | - | 3 | 316 | |
AK077948.1 | - | 3 | 308 | ||
AK151495.1 | - | 3 | 311 | ||
AK154811.1 | - | 3 | 323 | ||
BC115797.1 | - | 3 | 306 | 40103840 |