Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030112.2 | NP_084388.2 | 16, 17 | 1926 | |
GenBank mRNA | AK135633.1 | - | 10, 11 | 994 | |
BC117848.1 | - | 16, 17 | 1778 | 40093717 | |
BC117849.1 | - | 16, 17 | 1778 | 40093724 | |
CN532066.1 | - | 6, 7 | 442 |