Gene Transcripts

Gene Symbol
Adam26a
Entrez Gene ID
290763
Gene Name
a disintegrin and metallopeptidase domain 26A
Gene Aliases
Chromosome Location
Chr. 16: 51792060 - 51828519 on Build Rnor_6.0
UniGene ID
Rn.115533
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169119.1 NP_001162590.1 1254
GenBank mRNA BC085958.1 - 1254

View Full Product Details