Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006246834.2 | XP_006246896.1 | 11 | 1278 | |
XM_006246835.2 | XP_006246897.1 | 11 | 1278 | ||
XM_008767895.1 | XP_008766117.1 | 11 | 1278 | ||
XM_008767896.1 | XP_008766118.1 | 11 | 1278 |