Gene Transcripts

Gene Symbol
Aldh8a1
Entrez Gene ID
685750
Gene Name
aldehyde dehydrogenase 8 family, member A1
Gene Aliases
Chromosome Location
Chr. 1: 16910055 - 16929465 on Build Rnor_6.0
UniGene ID
Rn.44523
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191088.1 NP_001178017.1 3 371
XM_006227674.2 XP_006227736.1 3 406

View Full Product Details