Gene Transcripts

Gene Symbol
Peo1
Entrez Gene ID
309441
Gene Name
progressive external ophthalmoplegia 1
Gene Aliases
Chromosome Location
Chr. 1: 264756125 - 264762892 on Build Rnor_6.0
UniGene ID
Rn.105678
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107599.1 NP_001101069.1 1 1444
XM_008760429.1 XP_008758651.1 1 1504

View Full Product Details