Gene Transcripts

Gene Symbol
Arid4a
Entrez Gene ID
314205
Gene Name
AT rich interactive domain 4A (Rbp1 like)
Gene Aliases
Chromosome Location
Chr. 6: 93461713 - 93532901 on Build Rnor_6.0
UniGene ID
Rn.106963
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108029.1 NP_001101499.1 9, 10 1015
GenBank mRNA FQ233288.1 - 11, 12 1092

View Full Product Details