Gene Transcripts

Gene Symbol
Oxsr1
Entrez Gene ID
316064
Gene Name
oxidative-stress responsive 1
Gene Aliases
Osr1
Chromosome Location
Chr. 8: 127920349 - 128009951 on Build Rnor_6.0
UniGene ID
Rn.203085
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108194.1 NP_001101664.1 11 1352

View Full Product Details