Gene Transcripts

Gene Symbol
Akap17a
Entrez Gene ID
288526
Gene Name
A kinase (PRKA) anchor protein 17A
Gene Aliases
Chromosome Location
Chr. 12: 18527116 - 18531780 on Build Rnor_6.0
UniGene ID
Rn.24876
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127246.1 NP_001120718.1 5 1945
XM_006248998.2 XP_006249060.1 5 2658
XM_006248999.2 XP_006249061.1 5 2502
XM_006249000.2 XP_006249062.1 5 2593
GenBank mRNA BC166722.1 - 5 1945

View Full Product Details