Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013057.1 | NP_001013075.1 | 8 | 905 | |
XM_006254183.2 | XP_006254245.1 | 8 | 917 | ||
XM_006254184.2 | XP_006254246.1 | 8 | 896 | ||
XM_006254185.2 | XP_006254247.1 | 7 | 839 | ||
GenBank mRNA | BC078957.1 | - | 8 | 905 |