Gene Transcripts

Gene Symbol
Spns2
Entrez Gene ID
100270678
Gene Name
spinster homolog 2
Gene Aliases
Chromosome Location
Chr. 10: 59010428 - 59049482 on Build Rnor_6.0
UniGene ID
Rn.159761
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144991.1 NP_001138463.1 1 511
XM_006246562.2 XP_006246624.2 1 508
XM_006246563.2 XP_006246625.2 1 508

View Full Product Details