Gene Transcripts

Gene Symbol
Prl8a3
Entrez Gene ID
100361884
Gene Name
prolactin family 8, subfamily a, member 3
Gene Aliases
Plp-Cv, Plpc, Plpcv, Prlpc, Prlpc1
Chromosome Location
Chr. 17: 39455865 - 39463715 on Build Rnor_6.0
UniGene ID
Rn.202972
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020079.1 NP_064464.1 317
GenBank mRNA U93351.1 - 317

View Full Product Details