Gene Transcripts

Gene Symbol
Trg
Entrez Gene ID
24821
Gene Name
T cell receptor gamma locus
Gene Aliases
Tcrg, Trg@
Chromosome Location
Chr. : - on Build
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA D55648.1 - 3 795

View Full Product Details