Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001047853.1 | NP_001041318.1 | 3, 4 | 322 | |
XM_006229367.2 | XP_006229429.1 | 4, 5 | 864 | ||
XM_006229368.2 | XP_006229430.1 | 4, 5 | 864 | ||
GenBank mRNA | AY325151.1 | - | 3, 4 | 322 | |
AY539868.1 | - | 2, 3 | 282 | ||
FQ222682.1 | - | 4, 5 | 744 |