Gene Transcripts

Gene Symbol
Ccdc32
Entrez Gene ID
296081
Gene Name
coiled-coil domain containing 32
Gene Aliases
Gm631
Chromosome Location
Chr. 3: 110816327 - 110828824 on Build Rnor_6.0
UniGene ID
Rn.154291
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024245.1 NP_001019416.1 1 135
GenBank mRNA BC093612.1 - 1 135

View Full Product Details