Gene Transcripts

Gene Symbol
Prrt4
Entrez Gene ID
500059
Gene Name
proline-rich transmembrane protein 4
Gene Aliases
RGD1559885
Chromosome Location
Chr. 4: 56442599 - 56473787 on Build Rnor_6.0
UniGene ID
Rn.229283
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109226.1 NP_001102696.1 5 1745
XM_006236220.2 XP_006236282.1 5 1952
XM_008762817.1 XP_008761039.1 5 1636

View Full Product Details