Gene Transcripts

Gene Symbol
Arhgap25
Entrez Gene ID
500246
Gene Name
Rho GTPase activating protein 25
Gene Aliases
RGD1562105
Chromosome Location
Chr. 4: 119249682 - 119327822 on Build Rnor_6.0
UniGene ID
Rn.22776
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109247.1 NP_001102717.1 10 1995
XM_006236838.2 XP_006236900.1 10 4154
XM_006236839.2 XP_006236901.1 9 1850

View Full Product Details