Gene Transcripts

Gene Symbol
MAPK14
Entrez Gene ID
1432
Gene Name
mitogen-activated protein kinase 14
Gene Aliases
CSBP, CSBP1, CSBP2, CSPB1, EXIP, Mxi2, PRKM14, PRKM15, RK, SAPK2A, p38, p38ALPHA
Chromosome Location
Chr. 6: 36027635 - 36122964 on Build GRCh38
UniGene ID
Hs.485233
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001315.2 NP_001306.1 4, 5 887
NM_139012.2 NP_620581.1 4, 5 887
NM_139013.2 NP_620582.1 4, 5 887
NM_139014.2 NP_620583.1 4, 5 887
XM_006714998.3 XP_006715061.1 4, 5 362
XM_011514310.2 XP_011512612.2 4, 5 1155
XM_017010299.1 XP_016865788.1 4, 5 929
XM_017010300.1 XP_016865789.1 4, 5 929
XM_017010301.1 XP_016865790.1 4, 5 929
XM_017010302.1 XP_016865791.1 4, 5 929
XM_017010303.1 XP_016865792.1 4, 5 929
XM_017010304.1 XP_016865793.1 4, 5 929
GenBank mRNA AB074150.1 - 4, 5 406
AF100544.1 - 4, 5 421
AK291709.1 - 4, 5 782
AK303414.1 - 5, 6 451
BC000092.1 - 4, 5 744 3505234
BC031574.1 - 4, 5 743 5181064
BT006933.1 - 4, 5 406
CR536505.1 - 4, 5 406
FJ032367.1 - 4, 5 406
FJ032368.1 - 4, 5 406
L35263.1 - 4, 5 768
L35264.1 - 4, 5 614

View Full Product Details