Gene Transcripts
- Gene Symbol
- Nctc1
- Entrez Gene ID
- 330677
- Gene Name
- non-coding transcript 1
- Gene Aliases
- 9530074L01, Rhit1
- Chromosome Location
- Chr. 7: 142544609 - 142558598 on Build GRCm38
- UniGene ID
- Mm.265130
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002452.2 |
|
2 |
771 |
|
GenBank mRNA |
AK035596.1 |
- |
2 |
770 |
|
AK036720.1 |
- |
2 |
771 |
|
View Full Product Details