Gene Transcripts

Gene Symbol
Bambi-ps1
Entrez Gene ID
81913
Gene Name
BMP and activin membrane-bound inhibitor, pseudogene (Xenopus laevis)
Gene Aliases
Chromosome Location
Chr. 2: 122466583 - 122467797 on Build GRCm38
UniGene ID
Mm.122640
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027919.1 1 925
GenBank mRNA AK040009.1 - 1 869
AK041875.1 - 1 871
AK088217.1 - 1 864

View Full Product Details