Gene Transcripts

Gene Symbol
PSORS1C3
Entrez Gene ID
100130889
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
Gene Aliases
NCRNA00196
Chromosome Location
Chr. 6: 31173735 - 31177899 on Build GRCh38
UniGene ID
Hs.670091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026816.1 1 69
GenBank mRNA AB932952.1 - 1 69
AB932953.1 - 1 69
AB932954.1 - 1 69
AB933312.1 - 1 69
AB933313.1 - 1 69
AB933320.1 - 1 69
AY484516.1 - 1 69
LC027944.1 - 1 69
LC027945.1 - 2 361
LC027946.1 - 2 361
LC050979.1 - 2 361
LC050980.1 - 2 361
LC050981.1 - 2 361
LC050982.1 - 2 361
LC050983.1 - 2 361
LC050985.1 - 2 361
LC050986.1 - 2 361
LC050987.1 - 2 361
LC050989.1 - 2 361
LC050990.1 - 2 361
LC050991.1 - 2 361

View Full Product Details