Gene Transcripts

Gene Symbol
ANKRD13D
Entrez Gene ID
338692
Gene Name
ankyrin repeat domain 13D
Gene Aliases
Chromosome Location
Chr. 11: 67289291 - 67302484 on Build GRCh38
UniGene ID
Hs.438673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_030767.1 1, 2 214
XM_011544986.1 XP_011543288.1 1, 2 214
GenBank mRNA AK124721.1 - 1, 2 211
DA870287.1 - 1, 2 214

View Full Product Details