Gene Transcripts
- Gene Symbol
- PISRT1
- Entrez Gene ID
- 140464
- Gene Name
- polled intersex syndrome regulated transcript 1 (non-protein coding RNA)
- Gene Aliases
- NCRNA00195
- Chromosome Location
- Chr. 3: 139232992 - 139233522 on Build GRCh38
- UniGene ID
- Hs.721270
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027070.1 |
|
1 |
191 |
|
GenBank mRNA |
AI204197.1 |
- |
1 |
191 |
|
AW268472.1 |
- |
1 |
191 |
|
View Full Product Details