Gene Transcripts

Gene Symbol
AIFM3
Entrez Gene ID
150209
Gene Name
apoptosis inducing factor, mitochondria associated 3
Gene Aliases
AIFL
Chromosome Location
Chr. 22: 20965130 - 20981360 on Build GRCh38
UniGene ID
Hs.723246
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027464.1 3, 4 530
GenBank mRNA AK295581.1 - 3, 4 433
AK316042.1 - 3, 4 426


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DC316402.1 - 3, 4 438 Chr. 22 - 20967263 - 20974169 [+]
GenBankmRNA DC336866.1 - 3, 4 426 Chr. 22 - 20967263 - 20974276 [+]
GenBankmRNA DC314486.1 - 3, 4 427 Chr. 22 - 20967263 - 20974297 [+]
GenBankmRNA BP219618.1 - 3, 4 434 Chr. 22 - 20967255 - 20974290 [+]
GenBankmRNA DC314682.1 - 3, 4 434 Chr. 22 - 20967255 - 20974141 [+]
GenBankmRNA DC329777.1 - 3, 4 427 Chr. 22 - 20967263 - 20974273 [+]
GenBankmRNA DC326583.1 - 3, 4 429 Chr. 22 - 20967260 - 20974271 [+]
GenBankmRNA DC331924.1 - 3, 4 433 Chr. 22 - 20967256 - 20974297 [+]
GenBankmRNA DC331228.1 - 3, 4 438 Chr. 22 - 20967253 - 20974280 [+]
GenBankmRNA DC313174.1 - 3, 4 440 Chr. 22 - 20967249 - 20974141 [+]
GenBankmRNA DC316411.1 - 3, 4 434 Chr. 22 - 20967255 - 20974267 [+]
GenBankmRNA DC411284.1 - 3, 4 421 Chr. 22 - 20965172 - 20974175 [+]
GenBankmRNA DC313643.1 - 3, 4 429 Chr. 22 - 20967260 - 20974141 [+]
GenBankmRNA DA275204.1 - 3, 4 234 Chr. 22 - 20967277 - 20974774 [+]

View Full Product Details