Gene Transcripts

Gene Symbol
LINC01550
Entrez Gene ID
388011
Gene Name
long intergenic non-protein coding RNA 1550
Gene Aliases
C14orf64
Chromosome Location
Chr. 14: 97925610 - 97978124 on Build GRCh38
UniGene ID
Hs.651477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015430.1 2 258
GenBank mRNA AK091668.1 - 2 244
AK097943.1 - 2 113
BC043585.1 - 2 258
BX647564.1 - 2 242
JX073283.1 - 2 194


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA521036.1 - 2 244 Chr. 14 - 97960263 - 97978110 [-]
GenBankmRNA BI772351.1 - 2 182 Chr. 14 - 97959933 - 97978046 [-]
GenBankmRNA BX109572.1 - 2 203 Chr. 14 - 97968300 - 97978070 [-]
GenBankmRNA DB154307.1 - 2 199 Chr. 14 - 97968842 - 97978065 [-]
GenBankmRNA BI834723.1 - 2 261 Chr. 14 - 97926352 - 97978124 [-]
GenBankmRNA AA418794.1 - 2 171 Chr. 14 - 97968315 - 97978037 [-]

View Full Product Details