Gene Transcripts

Gene Symbol
ENSG00000186594
Entrez Gene ID
ENSG00000186594
Gene Name
Gene Aliases
Chromosome Location
Chr. : - on Build
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID

Gene Symbol
MIR22HG
Entrez Gene ID
84981
Gene Name
MIR22 host gene
Gene Aliases
C17orf91
Chromosome Location
Chr. 17: 1711504 - 1716272 on Build GRCh38
UniGene ID
Hs.597755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028502.1 2 343
NR_028503.1 2 343
NR_028504.1 3 426
NR_028505.1 2 343
GenBank mRNA BC119720.1 - 3 334
BC119721.1 - 3 334 40118088
BM755383.1 - 2 282
BX509423.1 - 2 280
BX640650.1 - 3 364
BX648321.1 - 2 280

View Full Product Details