Gene Transcripts

Gene Symbol
MIR22HG
Entrez Gene ID
84981
Gene Name
MIR22 host gene
Gene Aliases
C17orf91
Chromosome Location
Chr. 17: 1711504 - 1716272 on Build GRCh38
UniGene ID
Hs.597755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028502.1 2 2370
NR_028503.1 3 1563
NR_028504.1 4 1150
NR_028505.1 3 1067
GenBank mRNA AF070569.1 - 1 1939
BX640650.1 - 4 1088
BX648321.1 - 2 2307

View Full Product Details