Gene Transcripts
- Gene Symbol
- C3P1
- Entrez Gene ID
- 388503
- Gene Name
- complement component 3 precursor pseudogene
- Gene Aliases
- CPLP
- Chromosome Location
- Chr. 19: 10041356 - 10074138 on Build GRCh38
- UniGene ID
- Hs.655363
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027300.2 |
|
1 |
83 |
|
View Full Product Details