Gene Transcripts

Gene Symbol
MOCS1
Entrez Gene ID
4337
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
MIG11, MOCOD, MOCODA
Chromosome Location
Chr. 6: 39904258 - 39934551 on Build GRCh38
UniGene ID
Hs.357128
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033233.1 8, 9 1104
XM_011514633.2 XP_011512935.1 9, 10 1142
GenBank mRNA AF214023.1 - 3, 4 251
AK056740.1 - 8, 9 1050

View Full Product Details