Gene Transcripts
- Gene Symbol
- PQLC2L
- Entrez Gene ID
- 152078
- Gene Name
- PQ loop repeat containing 2 like
- Gene Aliases
- C3orf55
- Chromosome Location
- Chr. 3: 157543344 - 157601233 on Build GRCh38
- UniGene ID
- Hs.259046
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM716256.1 |
- |
2, 3 |
244 |
|
Chr. 3 - 157543385 - 157569529 [+] |
GenBankmRNA |
AI348631.1 |
- |
2, 3 |
241 |
|
Chr. 3 - 157543388 - 157569486 [+] |
GenBankmRNA |
BM709511.1 |
- |
2, 3 |
244 |
|
Chr. 3 - 157543385 - 157569663 [+] |
GenBankmRNA |
BG197365.1 |
- |
|
119 |
|
Chr. - - [-] |
GenBankmRNA |
BG200482.1 |
- |
1, 2 |
119 |
|
Chr. 3 - 157553230 - 157569865 [+] |
View Full Product Details