Gene Transcripts
- Gene Symbol
- MDS2
- Entrez Gene ID
- 259283
- Gene Name
- myelodysplastic syndrome 2 translocation associated
- Gene Aliases
- Chromosome Location
- Chr. 1: 23627334 - 23640566 on Build GRCh38
- UniGene ID
- Hs.523369
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027042.1 |
|
5 |
651 |
|
GenBank mRNA |
AJ310434.2 |
- |
5 |
651 |
|
BC069164.1 |
- |
5 |
651 |
|
View Full Product Details