Gene Transcripts
- Gene Symbol
- BTN2A3P
- Entrez Gene ID
- 54718
- Gene Name
- butyrophilin subfamily 2 member A3, pseudogene
- Gene Aliases
- BTN2.3, BTN2A3
- Chromosome Location
- Chr. 6: 26421391 - 26430588 on Build GRCh38
- UniGene ID
- Hs.729655
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027795.1 |
|
7 |
2047 |
|
GenBank mRNA |
AK056871.1 |
- |
7 |
2027 |
|
View Full Product Details