Gene Transcripts

Gene Symbol
LINC01554
Entrez Gene ID
202299
Gene Name
long intergenic non-protein coding RNA 1554
Gene Aliases
C5orf27, FIS
Chromosome Location
Chr. 5: 95852232 - 95860133 on Build GRCh38
UniGene ID
Hs.8373
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026936.1 3 1879
GenBank mRNA AK096140.1 - 4 1781
AY168789.1 - 4 1761


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000620167.1 - 1405 Chr. - - [-]

View Full Product Details