Gene Transcripts

Gene Symbol
LINC00488
Entrez Gene ID
677779
Gene Name
long intergenic non-protein coding RNA 488
Gene Aliases
C3orf66
Chromosome Location
Chr. 3: 109178165 - 109185261 on Build GRCh38
UniGene ID
Hs.618669
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026767.1 3 745
GenBank mRNA AK123226.1 - 3 745
BC022563.1 - 3 735


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI763241.1 - 3 719 Chr. 3 - 109178190 - 109183513 [+]

View Full Product Details