Gene Transcripts
- Gene Symbol
- CRHR1-IT1
- Entrez Gene ID
- 147081
- Gene Name
- CRHR1 intronic transcript 1
- Gene Aliases
- C17orf69
- Chromosome Location
- Chr. 17: 45638975 - 45646229 on Build GRCh38
- UniGene ID
- Hs.128813
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026905.2 |
|
1 |
509 |
|
NR_026906.2 |
|
1 |
509 |
|
GenBank mRNA |
BC069230.1 |
- |
1 |
230 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BF311196.1 |
- |
|
229 |
|
Chr. - - [-] |
GenBankmRNA |
BG534719.1 |
- |
1 |
511 |
|
Chr. 17 - 45638975 - 45646008 [+] |
GenBankmRNA |
BF312185.1 |
- |
1 |
227 |
|
Chr. 17 - 45639269 - 45640794 [+] |
GenBankmRNA |
BG187386.1 |
- |
1 |
111 |
|
Chr. 17 - 45639374 - 45646232 [+] |
GenBankmRNA |
CN283298.1 |
- |
1 |
78 |
|
Chr. 17 - 45639408 - 45639857 [+] |
View Full Product Details