Gene Transcripts

A Gene Symbol is not available because the non-coding siRNA was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID. Non-coding siRNAs are designed to RefSeq (NR_) or to curated sequences from RNAdb represented by GenBank RNA or ESTdb accessions.


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA U25472.1 - 1 18 Chr. 18 - 64150092 - 64150188 [-]
GenBankmRNA U25471.1 - 1 18 Chr. 6 - 109075378 - 109076142 [-]
GenBankmRNA U25462.1 - 1 18 Chr. 18 - 64150092 - 64150188 [-]

View Full Product Details