Gene Transcripts
- Gene Symbol
- RBM6
- Entrez Gene ID
- 10180
- Gene Name
- RNA binding motif protein 6
- Gene Aliases
- 3G2, DEF-3, DEF3, HLC-11, NY-LU-12, g16
- Chromosome Location
- Chr. 3: 49940038 - 50077252 on Build GRCh38
- UniGene ID
- Hs.696735
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124030.1 |
- |
1 |
127 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA173932.1 |
- |
1 |
324 |
|
Chr. 3 - 50067493 - 50068067 [+] |
GenBankmRNA |
DA958615.1 |
- |
1 |
127 |
|
Chr. 3 - 50067689 - 50068247 [+] |
GenBankmRNA |
BM908060.1 |
- |
1 |
227 |
|
Chr. 3 - 50067601 - 50068550 [+] |
View Full Product Details