Gene Transcripts

Gene Symbol
CEP83
Entrez Gene ID
51134
Gene Name
centrosomal protein 83
Gene Aliases
CCDC41, NPHP18, NY-REN-58
Chromosome Location
Chr. 12: 94268832 - 94459991 on Build GRCh38
UniGene ID
Hs.279209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006719437.2 XP_006719500.1 17 4757
XM_017019385.1 XP_016874874.1 18 4838
GenBank mRNA AK093735.1 - 1 1808

View Full Product Details