Gene Transcripts
- Gene Symbol
- ARHGEF6
- Entrez Gene ID
- 9459
- Gene Name
- Rac/Cdc42 guanine nucleotide exchange factor 6
- Gene Aliases
- COOL2, Cool-2, MRX46, PIXA, alpha-PIX, alphaPIX
- Chromosome Location
- Chr. X: 136665547 - 136781656 on Build GRCh38
- UniGene ID
- Hs.522795
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF207831.1 |
- |
1 |
341 |
|
View Full Product Details