Gene Transcripts
- Gene Symbol
- SLC15A4
- Entrez Gene ID
- 121260
- Gene Name
- solute carrier family 15 member 4
- Gene Aliases
- FP12591, PHT1, PTR4
- Chromosome Location
- Chr. 12: 128793194 - 128823996 on Build GRCh38
- UniGene ID
- Hs.507260
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097154.1 |
- |
1 |
848 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR936821.1 |
- |
1 |
4387 |
|
Chr. 12 - 128793757 - 128798825 [-] |
View Full Product Details