Gene Transcripts
- Gene Symbol
- PRRC2C
- Entrez Gene ID
- 23215
- Gene Name
- proline rich coiled-coil 2C
- Gene Aliases
- BAT2-iso, BAT2D1, BAT2L2, XTP2
- Chromosome Location
- Chr. 1: 171485496 - 171593511 on Build GRCh38
- UniGene ID
- Hs.494614
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124374.1 |
- |
1 |
537 |
|
View Full Product Details