Gene Transcripts
- Gene Symbol
- AHSA2
- Entrez Gene ID
- 130872
- Gene Name
- AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
- Gene Aliases
- Hch1
- Chromosome Location
- Chr. 2: 61177418 - 61191210 on Build GRCh38
- UniGene ID
- Hs.655602
- Species
- Homo sapiens
- Gene Symbol
- USP34
- Entrez Gene ID
- 9736
- Gene Name
- ubiquitin specific peptidase 34
- Gene Aliases
- Chromosome Location
- Chr. 2: 61187455 - 61470714 on Build GRCh38
- UniGene ID
- Hs.644708
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AW293998.1 |
- |
1 |
188 |
|
View Full Product Details