Gene Transcripts
- Gene Symbol
- KANSL1L
- Entrez Gene ID
- 151050
- Gene Name
- KAT8 regulatory NSL complex subunit 1 like
- Gene Aliases
- C2orf67, MSL1v2
- Chromosome Location
- Chr. 2: 210021421 - 210171370 on Build GRCh38
- UniGene ID
- Hs.282260
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX376230.2 |
- |
1 |
270 |
|
Chr. 2 - 210096310 - 210097005 [-] |
GenBankmRNA |
CR616459.1 |
- |
1 |
270 |
|
Chr. 2 - 210096310 - 210097005 [-] |
GenBankmRNA |
CD242070.1 |
- |
1 |
36 |
|
Chr. 2 - 210096305 - 210096768 [-] |
View Full Product Details