Gene Transcripts
- Gene Symbol
- AGBL3
- Entrez Gene ID
- 340351
- Gene Name
- ATP/GTP binding protein like 3
- Gene Aliases
- CCP3
- Chromosome Location
- Chr. 7: 134985169 - 135135782 on Build GRCh38
- UniGene ID
- Hs.648616
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021841.1 |
- |
1 |
666 |
|
AL832281.1 |
- |
1 |
2489 |
|
View Full Product Details