Gene Transcripts
- Gene Symbol
- HMCES
- Entrez Gene ID
- 56941
- Gene Name
- 5-hydroxymethylcytosine (hmC) binding, ES cell-specific
- Gene Aliases
- C3orf37, DC12, SRAPD1
- Chromosome Location
- Chr. 3: 129278806 - 129305293 on Build GRCh38
- UniGene ID
- Hs.458320
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF035296.1 |
- |
1 |
621 |
|
View Full Product Details