Gene Transcripts

Gene Symbol
C18orf12
Entrez Gene ID
84322
Gene Name
chromosome 18 open reading frame 12
Gene Aliases
HEIL1, HsT2508
Chromosome Location
Chr. 18: 48252301 - 48253185 on Build GRCh38
UniGene ID
Hs.334493
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017026109.1 XP_016881598.1 1 436
GenBank mRNA AB027121.1 - 1 436

View Full Product Details